EMAIL THIS PAGE TO A FRIEND
10305 Sigma-Aldrich

Bismuth(III) oxide

≥99.5% (complexometric)

Synonym: Dibismuth trioxide

Purchase

Properties

Related Categories Alternative Energy, Bismuth, Dopants, Materials Science, Metal and Ceramic Science,
InChI Key   WMWLMWRWZQELOS-UHFFFAOYSA-N
assay   ≥99.5% (complexometric)
loss   ≤0.5% loss on ignition, 1000 °C
anion traces   nitrate (NO3-): ≤500 mg/kg
cation traces   As: ≤5 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤100 mg/kg

Description

Features and Benefits

Preparation of new stabilized, oxide ion-conducting, bismuth vanadate phases by a microwave assisted method, from V2O5, Bi2O3 and other solid oxides, was reported. These ceramics show promise in solid oxide fuel cells, water-vapor electrolyzers and oxygen sensors.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

223891 Bismuth(III) oxide, ReagentPlus®, powder, 10 μm, 99.9% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?