• USA Home
  • 10611 - 12-(9-Anthracenecarbonyloxy)octadecanoic acid

EMAIL THIS PAGE TO A FRIEND
10611 Sigma-Aldrich

12-(9-Anthracenecarbonyloxy)octadecanoic acid

for fluorescence

Synonym: (±)-12-(9-Anthroyloxy)stearic acid, 12-(9-Anthracenecarbonyloxy)stearic acid

  • CAS Number 30536-60-8

  • Empirical Formula (Hill Notation) C33H44O4

  • Molecular Weight 504.70

  •  Beilstein Registry Number 1896752

  •  MDL number MFCD00056369

Purchase

Properties

Related Categories Alphabetical, Biochemicals and Reagents, Detection, Fluorescent Indicators and Probes, Fluorescent Probes, Labels, Particles and Stains,
grade   for fluorescence
InChI Key   KCSPZBFHFNHKIQ-UHFFFAOYSA-N
mp   76-78 °C(lit.)
solubility   DMF: soluble
  alcohols: soluble
suitability   in accordance for fluorescence
storage temp.   2-8°C

Description

Other Notes

Fluorescent fatty acid for studying lipid dynamics and lipid-protein interactions of biological membranes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?