EMAIL THIS PAGE TO A FRIEND
10781 Sigma-Aldrich

Antimony(III) oxide

puriss. p.a.

Synonym: Diantimony trioxide

Purchase

Properties

Related Categories A-B, Puriss p.a., Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Antimony,
InChI Key   ADCOVFLJGNWWNZ-UHFFFAOYSA-N
grade   puriss. p.a.
assay   ≥99.0% (RT)
bp   1550 °C(lit.)
mp   655 °C(lit.)
anion traces   chloride (Cl-): ≤500 mg/kg
  sulfate (SO42-): ≤100 mg/kg
cation traces   Ca: ≤20 mg/kg
  Cd: ≤20 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤20 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤50 mg/kg
  K: ≤20 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤20 mg/kg
  Ni: ≤10 mg/kg
  Pb: ≤250 mg/kg
  Zn: ≤20 mg/kg

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
2
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?