EMAIL THIS PAGE TO A FRIEND
10790 Sigma-Aldrich

Antipyrine

≥99.0% (RT)

Synonym: 2,3-Dimethyl-1-phenyl-3-pyrazolin-5-one, Phenazone

  • CAS Number 60-80-0

  • Empirical Formula (Hill Notation) C11H12N2O

  • Molecular Weight 188.23

  •  Beilstein Registry Number 157775

  •  EC Number 200-486-6

  •  MDL number MFCD00003146

  •  PubChem Substance ID 329749165

Purchase

Properties

Related Categories Building Blocks, C11, Chemical Synthesis, Heterocyclic Building Blocks, Pyrazoles More...
InChI Key   VEQOALNAAJBPNY-UHFFFAOYSA-N
assay   ≥99.0% (RT)
form   crystals
mp   109-111 °C(lit.)
  109-114 °C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
CD2450000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A5882 Antipyrine, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?