EMAIL THIS PAGE TO A FRIEND
10800 Sigma-Aldrich

Apiezon® K

oil (highly viscous)

Purchase

Properties

Related Categories Greases and Lubricants, Labware More...
form   oil (highly viscous)

Description

General description

with very low vapor pressure (10-3 at 300°C)

Legal Information

Apiezon is a registered trademark of M & I Materials Ltd.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?