EMAIL THIS PAGE TO A FRIEND
10801 Sigma-Aldrich

GC Stationary Phase

phase Apiezon L, tube of 50 g

Purchase

Properties

Related Categories Analytical/Chromatography, GC Stationary Phases, Gas Chromatography (GC), Packed GC Columns and Components
grade   stationary phase for GC
description   grease, free of additives
packaging   tube of 50 g
application(s)   gas chromatography (GC): suitable
matrix active group   Apiezon L phase
mp   47 °C(lit.)
transition temp   solidification point 30 °C

Description

Application

high vacuum grease

General description

Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?