• USA Home
  • 10861 - Succinylsulfathiazole

EMAIL THIS PAGE TO A FRIEND
10861 Sigma-Aldrich

Succinylsulfathiazole

~95% (TLC)

  • CAS Number 116-43-8

  • Empirical Formula (Hill Notation) C13H13N3O5S2

  • Molecular Weight 355.39

  •  Beilstein Registry Number 349989

  •  EC Number 204-141-0

  •  MDL number MFCD00022437

Purchase

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
WM4767000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

S8752 Succinylsulfathiazole, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?