• USA Home
  • 10872 - 3-Chloro-2,4-pentanedione

EMAIL THIS PAGE TO A FRIEND
10872 Sigma-Aldrich

3-Chloro-2,4-pentanedione

produced by Wacker Chemie AG, Burghausen, Germany, ≥97.0% (GC)

Synonym: 3-Chloroacetylacetone, 3-Cl-ACAC

  • CAS Number 1694-29-7

  • Linear Formula CH3COCHClCOCH3

  • Molecular Weight 134.56

  •  Beilstein Registry Number 605870

  •  EC Number 216-902-4

  •  MDL number MFCD00009651

  •  PubChem Substance ID 24846820

Purchase

Properties

Related Categories Building Blocks, C3 to C6, Carbonyl Compounds, Chemical Synthesis, Ketones,
grade   produced by Wacker Chemie AG, Burghausen, Germany
InChI Key   VLRGXXKFHVJQOL-UHFFFAOYSA-N
assay   ≥97.0% (GC)
mfr. no.   Wacker Chemie AG
refractive index   n20/D 1.483(lit.)
bp   49-52 °C/18 mmHg(lit.)
density   1.1921 g/mL at 20 °C(lit.)

Description

Other Notes

prices for bulk quantities on request

Same product — same quality — now Sigma-Aldrich brand

General description

3-chloro-2,4-pentanedione undergoes catalytic reduction with electrogenerated cobalt(I)salen via one-electron transfer to form 2,4-pentanedion-3-ate. It forms bis(3-chloro-2,4-pentanedionato)copper complex with copper.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN1224 - class 3 - PG 3 - Ketones, liquid, n.o.s., HI: all
WGK Germany 
3
Flash Point(F) 
132.8 °F
Flash Point(C) 
56 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?