• USA Home
  • 10953 - Araldite® M hardener 964

EMAIL THIS PAGE TO A FRIEND
10953 Sigma-Aldrich

Araldite® M hardener 964

Purchase

Description

Application

It was used in a study to demonstrate fabrication and properties of polyaminde and graphene oxide coated with carbon fiber reinforced epoxy composites. It was used to study electrochemical detection of dopamine and ascorbic acid neurotransmitters focusing on miniaturization of a graphite-epoxy and synthetic zeolite-modified graphite-epoxy composite macroelectrode as a quasi-microelectrode. It was also used in production of microfluidic chips using extremely low cost chemicals and equipment which was validated by producing monodisperse polymeric microparticles for drug delivery and hydrogel microfibers for cell embedding.

General description

Araldite® is an epoxy resin commonly used for embedding. It is basically free from polymerization damage and the plastic does not sublime in the microscope, hence providing continual support and preservation of fine detail. It is composed of araldite M or CY 212, dodecenyl succinic anhydride, dibutyl phthalate and 2,4,6-tridimethylamine methyl phenol.

Legal Information

Araldite is a registered trademark of Huntsman Advanced Materials Inc.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?