• USA Home
  • 10972 - LR Gold embedding kit

EMAIL THIS PAGE TO A FRIEND
10972 Sigma-Aldrich

LR Gold embedding kit

for microscopy

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Embedding & Mounting Media Kits, Embedding and Mounting Media, Microscopy Reagents More...
grade   for microscopy
storage temp.   2-8°C

Description

Components

Composition:
62659 LR Gold resin, 500 ml
62660 LR Accelerator, 10 ml
62663 LR Catalyst, 10 g

Application

Embedding medium with low viscosity for low temperature embedding of unfixed 1-4 μm sections. Useful for enzyme histochemistry and immuno cytochemistry. LR Gold is non-toxic.

General description

The components can also be ordered separately under the product numbers indicated.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 3316 9
WGK Germany 
3
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?