Synonym: sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol
CAS Number 67-63-0
Linear Formula (CH3)2CHOH
Molecular Weight 60.10
Beilstein Registry Number 635639
EC Number 200-661-7
MDL number MFCD00011674
PubChem Substance ID 24846940
Popular Documents: Specification Sheet (PDF)
| Related Categories | 2-Propanol (Isopropanol), ReagentPlus, ReagentPlus Solvent Grade Products, ACS and Reagent Grade Solvents, Amber Glass Bottles, |
| grade | Laboratory Reagent |
| vapor density | 2.1 (vs air) |
| vapor pressure | 33 mmHg ( 20 °C) |
| 44 mmHg ( 25 °C) | |
| assay | ≥99.5% |
| autoignition temp. | 750 °F |
| expl. lim. | 2 % |
Live Chat and Frequently Asked Questions
are available for this Product.
1, 4 L in glass bottle
20 L in steel drum
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?