Synonym: Butyl ether
CAS Number 142-96-1
Linear Formula [CH3(CH2)3]2O
Molecular Weight 130.23
Beilstein Registry Number 1732752
EC Number 205-575-3
MDL number MFCD00009461
PubChem Substance ID 24846975
Popular Documents: Specification Sheet (PDF)
| Related Categories | ReagentPlus, ReagentPlus Solvent Grade Products, ACS and Reagent Grade Solvents, Amber Glass Bottles, Carbon Steel Cans with NPT Threads, |
| grade | ReagentPlus® |
| vapor density | 4.48 (vs air) |
| vapor pressure | 4.8 mmHg ( 20 °C) |
| assay | ≥99% |
| autoignition temp. | 365 °F |
| expl. lim. | 8.5 % |
| refractive index |
n |
1 L in glass bottle
20, 200 L in steel drum
ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?