EMAIL THIS PAGE TO A FRIEND
11155 Sigma-Aldrich

D-Asparagine

puriss., anhydrous, ≥99.0% (NT)

Synonym: (R)-2-Aminosuccinamic acid, (R)-2-Aminosuccinic acid 4-amide, D-Aspartic acid 4-amide

  • CAS Number 2058-58-4

  • Linear Formula H2NCOCH2CH(NH2)CO2H

  • Molecular Weight 132.12

  •  Beilstein Registry Number 1723526

  •  EC Number 218-163-3

  •  MDL number MFCD00008036

Purchase

Properties

Related Categories Amino Acids & Derivatives, Asparagine, Chemical Biology, Chemical Synthesis, Peptide Chemistry More...
InChI Key   DCXYFEDJOCDNAF-UWTATZPHSA-N
grade   puriss.
assay   ≥99.0% (NT)
optical activity   [α]20/D −35.0±0.5°, c = 5% in 5 M HCl
mp   280 °C (dec.)(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?