EMAIL THIS PAGE TO A FRIEND
11300 Sigma-Aldrich

Asp-Phe methyl ester

≥99.0% (HPLC)

Synonym: N-(L-α-Aspartyl)-L-phenylalanine methyl ester, Asp-Phe-OMe, Aspartame

  • CAS Number 22839-47-0

  • Linear Formula HOOCCH2CH(NH2)CONHCH(CH2C6H5)COOCH3

  • Molecular Weight 294.30

  •  Beilstein Registry Number 2223850

  •  EC Number 245-261-3

  •  MDL number MFCD00002724

  •  PubChem Substance ID 329749352

Purchase

Properties

InChI Key   IAOZJIPTCAWIRG-QWRGUYRKSA-N
assay   ≥99.0% (HPLC)
impurities   ≤4.5% water
mp   242-248 °C
storage temp.   2-8°C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
WM3407000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A5139 Asp-Phe methyl ester, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?