• USA Home
  • 11335 - Atroxin from Bothrops atrox venom

EMAIL THIS PAGE TO A FRIEND
11335 Sigma-Aldrich

Atroxin from Bothrops atrox venom

Synonym: Bothrops atrox venom

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Snake, Toxins and Venoms
storage temp.   2-8°C

Description

Legal Information

Atroxin is a trademark of Sigma-Aldrich Co. LLC

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3462 6.1 / PGI
WGK Germany 
3
RTECS 
UK9588000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?