EMAIL THIS PAGE TO A FRIEND
11366 Sigma-Aldrich

Avocado oil

meets analytical specification of DAC 03

Synonym: Avocado oleum

Purchase

Properties

quality   meets analytical specification of DAC 03
refractive index   n20/D 1.469
density   0.92 g/mL at 20 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
CL2680000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?