• USA Home
  • 11588 - 4,4′-Azobis(4-cyanovaleric acid)

EMAIL THIS PAGE TO A FRIEND
11588 Sigma-Aldrich

4,4′-Azobis(4-cyanovaleric acid)

purum, moistened with water, ≥97.0% (calc. based on dry substance, T)

Synonym: 4,4′-Azobis(4-cyanopentanoic acid), ABCVA, ACVA

  • CAS Number 2638-94-0

  • Linear Formula HOCOCH2CH2C(CH3)(CN)N=NC(CH3)(CN)CH2CH2COOH

  • Molecular Weight 280.28

  •  Beilstein Registry Number 1729856

  •  EC Number 220-135-0

  •  MDL number MFCD00002799

  •  PubChem Substance ID 24847247

Purchase

Properties

Related Categories Azo/Diazo Compounds, Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   VFXXTYGQYWRHJP-FOCLMDBBSA-N
grade   purum
assay   ≥97.0% (calc. based on dry substance, T)
quality   moistened with water
contains   ~15% water as stabilizer
mp   118-125 °C (dec.)(lit.)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

4,4′-Azobis(4-cyanovaleric acid) has been used in the preparation of polystyrene particles by polymerizing styrene in ethyl alcohol.

Biochem/physiol Actions

4,4′-Azobis(4-cyanovaleric acid) is an azo-initiator that induces lipid peroxidation of sunflower oil.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1325 4.1 / PGII
WGK Germany 
2
RTECS 
YV6190500

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?