• USA Home
  • 11633 - α,α′-Azodiisobutyramidine dihydrochloride

EMAIL THIS PAGE TO A FRIEND
11633 Sigma-Aldrich

α,α′-Azodiisobutyramidine dihydrochloride

purum, ≥98.0% (AT)

Synonym: 2,2′-Azobis(2-methylpropionamidine) dihydrochloride, AAPH

  • CAS Number 2997-92-4

  • Empirical Formula (Hill Notation) C8H18N6 · 2HCl

  • Molecular Weight 271.19

  •  Beilstein Registry Number 3718854

  •  EC Number 221-070-0

  •  MDL number MFCD00142725

  •  PubChem Substance ID 329749521

Purchase

Properties

Related Categories Azo/Diazo Compounds, Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   LXEKPEMOWBOYRF-QDBORUFSSA-N
grade   purum
assay   ≥98.0% (AT)
mp   ~178 °C (dec.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN3226 - class 4.1 - Self-reactive solid type D (2,2'-AZOBIS(2-AMIDINOPROPANE)-DIHYDROCHLORIDE)
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

440914 2,2′-Azobis(2-methylpropionamidine) dihydrochloride, granular, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?