• USA Home
  • 11722 - Barium standard for AAS

EMAIL THIS PAGE TO A FRIEND
11722 Sigma-Aldrich

Barium standard for AAS

ready-to-use, traceable to BAM, in H2O

Synonym: Barium chloride solution

Purchase

Properties

Related Categories AAS, Additional Standards, Analytical Standards, Analytical/Chromatography, Single Solution,
InChI Key   WDIHJSXYQDMJHN-UHFFFAOYSA-L
shelf life   limited shelf life, expiry date on the label
concentration   in H2O
  1.000 g/L Ba
application(s)   ICP/AAS: suitable
format   single component solution

Description

Preparation Note

prepared with BaCl2.2H2O and H2O

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3264 8 / PGIII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?