• USA Home
  • 11760 - Barium chloride dihydrate

EMAIL THIS PAGE TO A FRIEND
11760 Sigma-Aldrich

Barium chloride dihydrate

puriss. p.a., ACS reagent, ≥99.0% (KT)

Purchase

Properties

Related Categories A-B, Puriss p.a. ACS, Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Barium,
grade   ACS reagent
InChI Key   PWHCIQQGOQTFAE-UHFFFAOYSA-L
grade   puriss. p.a.
assay   ≥99.0% (KT)
impurities   ≤0.002% total nitrogen (N)
pH   5.2-8.2 (25 °C, 50 mg/mL in H2O)
cation traces   Ca: ≤50 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤1 mg/kg
  K: ≤25 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Sr: ≤1000 mg/kg
  Zn: ≤5 mg/kg

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1564 6.1 / PGIII
WGK Germany 
1
RTECS 
CQ8751000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?