• USA Home
  • 11770 - Barium citrate tribasic heptahydrate

EMAIL THIS PAGE TO A FRIEND
11770 Sigma-Aldrich

Barium citrate tribasic heptahydrate

puriss. p.a., ≥98.0%

Synonym: Citric acid barium salt, Tribarium dicitrate heptahydrate

  • CAS Number 6487-29-2

  • Linear Formula Ba3(C6H5O7)2 · 7H2O

  • Molecular Weight 916.29

  •  Beilstein Registry Number 3924511

  •  EC Number 208-140-6

  •  MDL number MFCD01862635

  •  PubChem Substance ID 329749550

Purchase

Properties

Related Categories A-B, Puriss p.a., Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Barium Salts,
InChI Key   LBYFZIZBHOKEJA-UHFFFAOYSA-H
grade   puriss. p.a.
assay   ≥98.0% (calc. on dry substance, KT)
  ≥98.0%
impurities   ≤0.01% insoluble matter in HCl
anion traces   chloride (Cl-): ≤10 mg/kg
  nitrate (NO3-): ≤20 mg/kg
cation traces   Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤10 mg/kg
  K: ≤50 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Sr: ≤5000 mg/kg
  Zn: ≤5 mg/kg

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?