EMAIL THIS PAGE TO A FRIEND
11848 Sigma-Aldrich

Barium titanate(IV)

purum, ≥98.0%

Synonym: Titanium barium oxide

Purchase

Properties

Related Categories Barium, Catalysis and Inorganic Chemistry, Ceramics, Ceramics by Element, Chemical Synthesis,
InChI Key   WNKMTAQXMLAYHX-UHFFFAOYSA-N
grade   purum
assay   ≥98.0%
form   powder
loss   ≤1% loss on ignition
particle size   <3 μm
density   6.08 g/mL at 25 °C(lit.)

Description

Analysis Note

ICP assay confirms Ba and Ti components

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
XR1437333

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?