• USA Home
  • 11919 - Buffer Standard solution pH 10.01

EMAIL THIS PAGE TO A FRIEND
11919 Sigma-Aldrich

Buffer Standard solution pH 10.01

ready-to-use, Na2CO3/NaHCO3, bag with 30 ml, traceable to NIST standards

Synonym: Sodium carbonate buffer solution

Purchase

Properties

quality   bag with 30 ml
  traceable to NIST standards
shelf life   limited shelf life, expiry date on the label
impurities   Na2CO3/NaHCO3

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?