• USA Home
  • 11921 - Buffer Standard solution pH 4.01

EMAIL THIS PAGE TO A FRIEND
11921 Sigma-Aldrich

Buffer Standard solution pH 4.01

ready-to-use, bag with 30 ml, traceable to NIST standards

Synonym: Potassium phthalate buffer solution

Purchase

Properties

quality   bag with 30 ml
  traceable to NIST standards
shelf life   limited shelf life, expiry date on the label
impurities   potassium hydrogen phthalate

Price and Availability

Safety & Documentation

Safety Information

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?