EMAIL THIS PAGE TO A FRIEND
12015 Sigma-Aldrich

Benzaldehyde

purum, ≥98.0% (GC)

Synonym: Bitter almond

  • CAS Number 100-52-7

  • Linear Formula C6H5CHO

  • Molecular Weight 106.12

  •  Beilstein Registry Number 471223

  •  EC Number 202-860-4

  •  MDL number MFCD00003299

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C7, Carbonyl Compounds, Chemical Synthesis,
vapor density   3.7 (vs air)
vapor pressure   4 mmHg ( 45 °C)
InChI Key   HUMNYLRZRPPJDN-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
autoignition temp.   374 °F
expl. lim.   1.4 %, 20 °F
impurities   chlorine, none detected
refractive index   n20/D 1.545(lit.)
  n20/D 1.545
bp   178-179 °C(lit.)
mp   −26 °C(lit.)
density   1.044 g/cm3 at 20 °C(lit.)

Description

Other Notes

Sales restrictions may apply

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1990 9 / PGIII
WGK Germany 
1
RTECS 
CU4375000
Flash Point(F) 
147.2 °F
Flash Point(C) 
64 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?