• USA Home
  • 12025 - Benzaldehyde dimethyl acetal

EMAIL THIS PAGE TO A FRIEND
12025 Sigma-Aldrich

Benzaldehyde dimethyl acetal

purum, ≥98.0% (GC)

Synonym: α,α-Dimethoxytoluene

  • CAS Number 1125-88-8

  • Linear Formula C6H5CH(OCH3)2

  • Molecular Weight 152.19

  •  Beilstein Registry Number 2044501

  •  EC Number 214-413-0

  •  MDL number MFCD00008491

  •  PubChem Substance ID 24847413

Purchase

Properties

Related Categories Acetals/Ketals/Ortho Esters, Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds More...
InChI Key   HEVMDQBCAHEHDY-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.493(lit.)
  n20/D 1.494
bp   87-89 °C/18 mmHg(lit.)
density   1.014 g/mL at 25 °C(lit.)

Description

Other Notes

Reagent for a convenient introduction of the O-benzylidene group into carbohydrates

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
RTECS 
CU5774000
Flash Point(F) 
156.2 °F
Flash Point(C) 
69 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?