• USA Home
  • 12103 - Tetraundecyl benzhydrol-3,3′,4,4′-tetracarboxylate

EMAIL THIS PAGE TO A FRIEND
12103 Sigma-Aldrich

Tetraundecyl benzhydrol-3,3′,4,4′-tetracarboxylate

Selectophore

Synonym: ETH 2112, Tetraundecyl α-hydroxy-4,4′-methylene-diphthalate

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Plasticizer for ISE More...
grade   Selectophore
InChI Key   ITKYCYSYWXAVCC-UHFFFAOYSA-N
mp   38-40 °C
storage temp.   2-8°C

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Plasticizer of high lipophilicity for clinically relevant ion-selective electrodes (ISE) and multisensing field effect transistors (ISFET)

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?