• USA Home
  • 12245 - N-Methyl-N-trimethylsilyltrifluoroacetamide activated I

EMAIL THIS PAGE TO A FRIEND
12245 Sigma-Aldrich

N-Methyl-N-trimethylsilyltrifluoroacetamide activated I

for GC, activated with ethanethiol and ammonium iodide

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Derivatization Reagents, Derivatization Reagents GC, Reagents for Silylation,
grade   for GC
refractive index   n20/D 1.380
storage temp.   2-8°C

Description

Analysis Note

applications test: suitable for the silylation of testosterone, tested by GCMS

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
78.8 °F
Flash Point(C) 
26 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

Silylating Reagents for GC-MS Compatible Derivatives of NSAIDs

Human and veterinary pharmaceuticals are a group of “emerging” contaminants,1, 2 some of which are produced in increasingly large volumes every year. The amounts produced are reaching quantities simi...
Luis-Alberto Martin
AnalytiX Volume 10 Article 2
Keywords: Anti-inflammatory agents, Derivatizations, Environmental, Gas chromatography, Gas chromatography mass spectrometry, Mass spectrometry, Metabolites, Pesticides, Pharmaceutical, Silylations, Veterinary

Related Products

related product

Product #

Description

Add to Cart

50992 N-Methyl-N-trimethylsilyltrifluoroacetamide activated I, for GC derivatization, activated with ethanethiol and ammonium iodide

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?