EMAIL THIS PAGE TO A FRIEND
123226 Sigma-Aldrich

Anisole

ReagentPlus®, 99%

Synonym: Methoxybenzene, Methyl phenyl ether

Purchase

Properties

Related Categories ReagentPlus, ReagentPlus Solvent Grade Products, ACS and Reagent Grade Solvents, Amber Glass Bottles, Carbon Steel Cans with NPT Threads,
grade   ReagentPlus®
vapor density   3.7 (vs air)
vapor pressure   10 mmHg ( 42.2 °C)
assay   99%
form   liquid
autoignition temp.   887 °F
refractive index   n20/D 1.516(lit.)
bp   154 °C(lit.)
mp   −37 °C(lit.)
density   0.995 g/mL at 25 °C(lit.)

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Packaging

1, 4 L in glass bottle

100 mL in glass bottle

18, 20 L in steel drum

Legal Information

ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN 2222 3 / PGIII
WGK Germany 
2
Flash Point(F) 
123.8 °F
Flash Point(C) 
51 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Determination of Water Content in Anisole Using Karl Fischer Titration

Ethers behave similarly to hydrocarbons. They are mostly adequately soluble in methanol. Only long chain ethers require the addition of propanol or chloroform as solubilizers. Double bonds can someti...
Keywords: Titrations

Fluorescence Quenching Microscopy: Imaging Two-Dimensional Materials

Alane Lim and Jiaxing Huang* Department of Materials Science and Engineering, Northwestern University, Evanston, IL 60208, USA *Email: jiaxing-huang@northwestern.edu
Keywords: Chemical vapor deposition, Deposition, Diffraction, Materials Science, Microscopy, Optical microscopy, Scanning electron microscopy, Solvents, Spectroscopy, Spin coating

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?