• USA Home
  • 12357 - Iron(III) sulfate hydrate

EMAIL THIS PAGE TO A FRIEND
12357 Sigma-Aldrich

Iron(III) sulfate hydrate

puriss., 21-23% Fe basis

Synonym: Ferric sulfate hydrate

Purchase

Properties

Related Categories Essential Chemicals, Iron Salts, Research Essentials, Salts
InChI Key   RUTXIHLAWFEWGM-UHFFFAOYSA-H
grade   puriss.
assay   21-23% Fe basis
anion traces   chloride (Cl-): ≤100 mg/kg
cation traces   Cu: ≤50 mg/kg
  Fe2+: ≤500 mg/kg
  Zn: ≤50 mg/kg

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Redi-Dri
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?