EMAIL THIS PAGE TO A FRIEND
12488 Sigma-Aldrich

Benzo[k]fluoranthene

puriss., for fluorescence, ≥99.0% (GC)

  • CAS Number 207-08-9

  • Empirical Formula (Hill Notation) C20H12

  • Molecular Weight 252.31

  •  Beilstein Registry Number 1873745

  •  EC Number 205-916-6

  •  MDL number MFCD00046287

  •  PubChem Substance ID 329749745

Purchase

Properties

grade   for fluorescence
InChI Key   HAXBIWFMXWRORI-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (GC)
mp   215-217 °C(lit.)
  215-217 °C
solubility   95% ethanol: <1 mg/mL at 20 °C
  DMSO: <1 mg/mL at 20 °C
  DMSO: <1 mg/mL at 20 °C
  H2O: <1 mg/mL at 20 °C
  acetone: 1-10 mg/mL at 20 °C
  methanol: <1 mg/mL at 20 °C
  toluene: 5-10 mg/mL at 20 °C
fluorescence   λex 306 nm; λem 411 nm in acetonitrile: water (1:1)

Description

Analysis Note

In addition to the emission maximum at 411 nm, there are lower maxima at 435 and 460 nm.

Application

Non-carcinogenic standard in environmental research

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
DF6350000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?