EMAIL THIS PAGE TO A FRIEND
12560 Sigma-Aldrich

Benzene

purum, ≥99.0% (GC)

  • CAS Number 71-43-2

  • Empirical Formula (Hill Notation) C6H6

  • Molecular Weight 78.11

  •  Beilstein Registry Number 969212

  •  EC Number 200-753-7

  •  MDL number MFCD00003009

  •  PubChem Substance ID 329749759

Purchase

Properties

Related Categories Benzene and Benzene Solutions, Solvent by Type, Solvents More...
vapor density   2.77 (vs air)
vapor pressure   166 mmHg ( 37.7 °C)
  74.6 mmHg ( 20 °C)
InChI Key   UHOVQNZJYSORNB-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
form   liquid
  liquid
autoignition temp.   1043 °F
expl. lim.   8 %
impurities   thiophene, none detected
refractive index   n20/D 1.501(lit.)
  n20/D 1.501
bp   80 °C(lit.)
mp   5.5 °C(lit.)
density   0.874 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN1114 - class 3 - PG 2 - Benzene
WGK Germany 
3
RTECS 
CY1400000
Flash Point(F) 
12.2 °F
Flash Point(C) 
-11 °C
Protocols & Articles

Related Content

Properties of Solvents Table

a. Gant, R., LC/GC 10(7), 515 (1992) 1. At 20 °C 2. At 20 °C +/- 5 °C relative to water at 4 °C 3. At 10mmHg pressure 4. At l=589.32nm
Keywords: Gas chromatography, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?