• USA Home
  • 12565 - Benzene-chromium(0) tricarbonyl

EMAIL THIS PAGE TO A FRIEND
12565 Sigma-Aldrich

Benzene-chromium(0) tricarbonyl

purum, ≥98.0% (C)

Synonym: Benchrotrene, Benzene-tricarbonylchromium(0)

  • CAS Number 12082-08-5

  • Linear Formula (C6H6)Cr(CO)3

  • Molecular Weight 214.14

  •  Beilstein Registry Number 3571699

  •  EC Number 235-146-6

  •  MDL number MFCD00009955

Purchase

Properties

InChI Key   WVSBQYMJNMJHIM-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (C)
mp   163-166 °C(lit.)

Description

Application

for synthesis of substituted benzene derivatives

Catalyst for:
• Oxidation of alkenes
• Hydrogenation of alkynes

Other Notes

Synthesis of substituted derivatives

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
GB4700000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

119318 Benzene-chromium(0) tricarbonyl, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?