• USA Home
  • 12585 - Benzeneseleninic acid

EMAIL THIS PAGE TO A FRIEND
12585 Sigma-Aldrich

Benzeneseleninic acid

purum, ≥98.0% (T)

  • CAS Number 6996-92-5

  • Linear Formula C6H5SeO2H

  • Molecular Weight 189.07

  •  Beilstein Registry Number 1929765

  •  EC Number 230-271-2

  •  MDL number MFCD00002100

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Selenium Compounds
InChI Key   WIHKGDVGLJJAMC-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (T)
mp   121-124 °C(lit.)
  122-124 °C

Description

Other Notes

Epoxidation reagent; Versatile oxidising reagent

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN3283 - class 6.1 - PG 3 - EHS - Selenium compound, solid, n.o.s., HI: all (not BR)
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

213004 Benzeneseleninic acid, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?