• USA Home
  • 12610 - Sodium benzenesulfinate

EMAIL THIS PAGE TO A FRIEND
12610 Sigma-Aldrich

Sodium benzenesulfinate

purum, ≥97.0% (HPLC)

Synonym: Benzenesulfinic acid sodium salt

  • CAS Number 873-55-2

  • Linear Formula C6H5SO2Na

  • Molecular Weight 164.16

  •  Beilstein Registry Number 3598405

  •  EC Number 212-842-8

  •  MDL number MFCD00013135

  •  PubChem Substance ID 24847699

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonic/Sulfinic Acid Salts, Sulfur Compounds More...
InChI Key   CHLCPTJLUJHDBO-UHFFFAOYSA-M
grade   purum
assay   ≥97.0% (HPLC)
impurities   ≤2% water

Description

Other Notes

This product has been replaced by 123579-ALDRICH | Benzenesulfinic acid sodium salt 98%

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?