• USA Home
  • 12643 - Potassium hexacyanoferrate(III)

EMAIL THIS PAGE TO A FRIEND
12643 Sigma-Aldrich

Potassium hexacyanoferrate(III)

purum, ≥99% (iodometric)

Synonym: Potassium ferricyanide, Red prussiate

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Essential Chemicals, Iron, Potassium Salts,
InChI Key   MIMJFNVDBPUTPB-UHFFFAOYSA-N
grade   purum
assay   ≥99% (iodometric)
form   crystalline
anion traces   chloride (Cl-): ≤1000 mg/kg
  sulfate (SO42-): ≤100 mg/kg

Description

Price and Availability


Redi-Dri
Safety & Documentation

Safety Information

Supplemental Hazard Statements 
Contact with acids liberates very toxic gas.
Personal Protective Equipment 
WGK Germany 
3
RTECS 
LJ8225000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?