• USA Home
  • 12680 - Sodium benzenesulfonate

EMAIL THIS PAGE TO A FRIEND
12680 Sigma-Aldrich

Sodium benzenesulfonate

puriss., recryst., ≥99.0% (T)

Synonym: Benzenesulfonic acid sodium salt

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonic/Sulfinic Acid Salts, Sulfur Compounds More...
InChI Key   MZSDGDXXBZSFTG-UHFFFAOYSA-M
grade   puriss.
assay   ≥99.0% (T)
quality   recryst.
impurities   ≤1% water
density   1.124 g/mL at 25 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DB7350000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

147281 Sodium benzenesulfonate, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?