• USA Home
  • 12690 - Sodium benzenesulfonate

EMAIL THIS PAGE TO A FRIEND
12690 Sigma-Aldrich

Sodium benzenesulfonate

purum, ≥98.0% (T)

Synonym: Benzenesulfonic acid sodium salt

  • CAS Number 515-42-4

  • Linear Formula C6H5SO3Na

  • Molecular Weight 180.16

  •  Beilstein Registry Number 3918459

  •  EC Number 208-198-2

  •  MDL number MFCD00065179

  •  PubChem Substance ID 24847750

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonic/Sulfinic Acid Salts, Sulfur Compounds More...
InChI Key   MZSDGDXXBZSFTG-UHFFFAOYSA-M
grade   purum
assay   ≥98.0% (T)
impurities   ≤7% water
density   1.124 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DB7350000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?