• USA Home
  • 12715 - Benzo[f]ninhydrin monohydrate

EMAIL THIS PAGE TO A FRIEND
12715 Sigma-Aldrich

Benzo[f]ninhydrin monohydrate

for fluorescence, ≥98.0% (HPLC)

Synonym: 2,2-Dihydroxy-(1H)-benz[f]indene-1,3(2H)-dione

  • CAS Number 38627-57-5

  • Empirical Formula (Hill Notation) C13H6O3 · H2O

  • Molecular Weight 228.20

  •  Beilstein Registry Number 2459087

  •  MDL number MFCD00012249

  •  PubChem Substance ID 24847765

Purchase

Description

Other Notes

Reagent for the chemical development of latent fingerprints

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?