EMAIL THIS PAGE TO A FRIEND
12740 Sigma-Aldrich

Benzo[ghi]perylene

purum, ≥98.0% (GC)

Synonym: 1,12-Benzoperylene

  • CAS Number 191-24-2

  • Empirical Formula (Hill Notation) C22H12

  • Molecular Weight 276.33

  •  Beilstein Registry Number 1913029

  •  EC Number 205-883-8

  •  MDL number MFCD00004135

  •  PubChem Substance ID 329749859

Purchase

Properties

Related Categories Arenes, Building Blocks, Chemical Synthesis, Organic Building Blocks
InChI Key   GYFAGKUZYNFMBN-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
bp   >500 °C(lit.)
mp   276-279 °C
  277-279 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?