EMAIL THIS PAGE TO A FRIEND
12799 Sigma-Aldrich

1H-Benzotriazole

≥98.0% (N)

Synonym: 1,2,3-Benzotriazole

  • CAS Number 95-14-7

  • Empirical Formula (Hill Notation) C6H5N3

  • Molecular Weight 119.12

  •  Beilstein Registry Number 112133

  •  EC Number 202-394-1

  •  MDL number MFCD00005699

  •  PubChem Substance ID 329749880

Purchase

Properties

Related Categories Benzotriazoles, Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks
InChI Key   QRUDEWIWKLJBPS-UHFFFAOYSA-N
assay   ≥98.0% (N)
ign. residue   ~0.5%
mp   94-99 °C

Description

Other Notes

Auxiliary used in the synthesis of amines from glyoxal

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
DM1225000
Flash Point(F) 
338 °F
Flash Point(C) 
170 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?