• USA Home
  • 12800 - α,α,α-Trichlorotoluene

EMAIL THIS PAGE TO A FRIEND
12800 Sigma-Aldrich

α,α,α-Trichlorotoluene

purum, ≥97.0% (GC)

Synonym: (Trichloromethyl)benzene, Benzotrichloride

Purchase

Properties

Related Categories Aryl, Building Blocks, C7, Chemical Synthesis, Halogenated Hydrocarbons,
vapor density   6.77 (vs air)
vapor pressure   0.2 mmHg ( 20 °C)
InChI Key   XEMRAKSQROQPBR-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
autoignition temp.   411 °F
refractive index   n20/D 1.557(lit.)
  n20/D 1.559
bp   219-223 °C/760 mmHg(lit.)
  90-91 °C/15 mmHg(lit.)
mp   −7.5-−7 °C(lit.)
density   1.38 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2226 8 / PGII
WGK Germany 
3
RTECS 
XT9275000
Flash Point(F) 
228.2 °F
Flash Point(C) 
109 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

147893 α,α,α-Trichlorotoluene, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?