• USA Home
  • 12914 - Cobalt(II) chloride hexahydrate

EMAIL THIS PAGE TO A FRIEND
12914 Sigma-Aldrich

Cobalt(II) chloride hexahydrate

puriss., ≥99%

Synonym: Cobaltous chloride hexahydrate

Purchase

Properties

Related Categories Cobalt Salts, Essential Chemicals, Research Essentials, Salts
vapor pressure   40 mmHg ( 0 °C)
InChI Key   GFHNAMRJFCEERV-UHFFFAOYSA-L
grade   puriss.
assay   ≥99%
anion traces   sulfate (SO42-): ≤50 mg/kg
cation traces   Ca: ≤500 mg/kg
  Fe: ≤50 mg/kg
  K: ≤100 mg/kg
  Mg: ≤100 mg/kg
  Na: ≤100 mg/kg
  Ni: ≤1000 mg/kg
  Pb: ≤20 mg/kg
  Zn: ≤500 mg/kg

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
GG0200000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?