• USA Home
  • 12927 - 4-Benzoylbenzoic acid

EMAIL THIS PAGE TO A FRIEND
12927 Sigma-Aldrich

4-Benzoylbenzoic acid

purum, ≥98.0% (T)

Synonym: Benzophenone-4-carboxylic acid

  • CAS Number 611-95-0

  • Linear Formula C6H5COC6H4CO2H

  • Molecular Weight 226.23

  •  Beilstein Registry Number 1960224

  •  EC Number 210-286-0

  •  MDL number MFCD00002560

  •  PubChem Substance ID 329749942

Purchase

Properties

Related Categories Building Blocks, C13 to C42+, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   IFQUPKAISSPFTE-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (T)
mp   198-200 °C(lit.)
  198-200 °C
Gene Information   human ... ACHE(43), BCHE(590), CES1(1066)

Description

Application

4-Benzoylbenzoic acid (Benzophenone-4-carboxylic acid) was used as a photoactive cross-linking reagent in the preparation of monoclonal antibodies. It was also used to graft a Gly-Arg-Gly-Asp-Ser-Pro (GRGDSP) peptide on the surface of the poly(carbonate urethane)s.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?