EMAIL THIS PAGE TO A FRIEND
13153 Sigma-Aldrich

6-Benzylaminopurine

≥99.0% (HPLC)

Synonym: 6-BAP, BA, N6-Benzyladenine

  • CAS Number 1214-39-7

  • Empirical Formula (Hill Notation) C12H11N5

  • Molecular Weight 225.25

  •  Beilstein Registry Number 19406

  •  EC Number 214-927-5

  •  MDL number MFCD00005572

  •  PubChem Substance ID 329750029

Purchase

Properties

InChI Key   NWBJYWHLCVSVIJ-UHFFFAOYSA-N
assay   ≥99.0% (HPLC)
mp   230-233 °C
anion traces   sulfate (SO42-): ≤150 mg/kg
cation traces   Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤50 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤5000 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg

Description

Analysis Note

plant cell culture tested

Other Notes

Plant cell division factor. Studies on growth regulators in tissue culture media in the presence of active charcoal.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
AU6252200

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?