EMAIL THIS PAGE TO A FRIEND
13280 Sigma-Aldrich

Benzyl chloride

purum, ≥99.0% (GC)

Synonym: α-Chlorotoluene

Purchase

Properties

Related Categories Aryl, Building Blocks, C7, Chemical Synthesis, Halogenated Hydrocarbons,
vapor density   4.36 (vs air)
vapor pressure   10.3 mmHg ( 60 °C)
  7 mmHg ( 55 °C)
InChI Key   KCXMKQUNVWSEMD-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
autoignition temp.   1085 °F
expl. lim.   14 %
refractive index   n20/D 1.538(lit.)
  n20/D 1.539
bp   177-181 °C(lit.)
mp   −43 °C(lit.)
solubility   : soluble0.46 g/L at 30 °C (Decomposes in contact with water)
density   1.1 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 1738 8(6.1) / PGII
WGK Germany 
3
RTECS 
XS8925000
Flash Point(F) 
152.6 °F
Flash Point(C) 
67 °C
Protocols & Articles

Articles

US EPA Method TO-17: GC Analysis of Volatiles on Equity®-1 after Collection/Desorption using Carbotrap 300 Tube

From our library of Articles, Sigma-Aldrich presents US EPA Method TO-17: GC Analysis of Volatiles on Equity®-1 after Collection/Desorption using Carbotrap 300 Tube
Keywords: Chromatography, Gas chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?