• USA Home
  • 13285 - N-Benzylcinchonidinium chloride

EMAIL THIS PAGE TO A FRIEND
13285 Sigma-Aldrich

N-Benzylcinchonidinium chloride

purum, ≥98.0% (AT)

Synonym: BCDC

  • CAS Number 69257-04-1

  • Empirical Formula (Hill Notation) C26H29ClN2O

  • Molecular Weight 420.97

  •  Beilstein Registry Number 5421832

  •  EC Number 273-938-3

  •  MDL number MFCD00082422

  •  PubChem Substance ID 329750126

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Cinchona Alkaloids, Privileged Ligands and Complexes More...
InChI Key   FCHYSBWCOKEPNQ-IOPLZPHGSA-M
grade   purum
assay   ≥98.0% (AT)
optical activity   [α]20/D −188±3°, c = 0.4% in H2O

Description

Other Notes

Chiral phase transfer catalyst ; resolutions by crystallization of the inclusion complex

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?