• USA Home
  • 13375 - Benzyldodecyldimethylammonium bromide

EMAIL THIS PAGE TO A FRIEND
13375 Sigma-Aldrich

Benzyldodecyldimethylammonium bromide

purum, ≥97.0% (AT)

Synonym: Benzyldimethyldodecylammonium bromide

  • CAS Number 7281-04-1

  • Linear Formula CH3(CH2)11N(Br)(CH3)2CH2C6H5

  • Molecular Weight 384.44

  •  Beilstein Registry Number 3579193

  •  EC Number 230-698-4

  •  MDL number MFCD00011768

  •  PubChem Substance ID 329750154

Purchase

Properties

Related Categories Ammonium Salts, Biochemicals and Reagents, Cationic Detergents, Chemical Synthesis, Detergents,
description   cationic
InChI Key   KHSLHYAUZSPBIU-UHFFFAOYSA-M
grade   purum
assay   ≥97.0% (AT)
mp   50-55 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
2
RTECS 
BO7875000
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

13373 Benzyldodecyldimethylammonium bromide, ≥99.0% (AT)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?