• USA Home
  • 13400 - Benzyldimethylhexadecylammonium chloride

EMAIL THIS PAGE TO A FRIEND
13400 Sigma-Aldrich

Benzyldimethylhexadecylammonium chloride

≥97.0% (dried material, AT)

Synonym: 16-BAC, Benzylhexadecyldimethylammonium chloride, Cetalkonium chloride, Cetyldimethylbenzylammonium chloride

  • CAS Number 122-18-9

  • Linear Formula CH3(CH2)15N(Cl)(CH3)2CH2C6H5

  • Molecular Weight 396.09

  •  Beilstein Registry Number 3802341

  •  EC Number 204-526-3

  •  MDL number MFCD00011625

  •  PubChem Substance ID 329750161

Purchase

Properties

Related Categories Ammonium Salts, Biochemicals and Reagents, Cationic Detergents, Chemical Synthesis, Detergents,
description   cationic
InChI Key   SXPWTBGAZSPLHA-UHFFFAOYSA-M
assay   ≥97.0% (dried material, AT)
impurities   ≤9% water
mp   55-65 °C(lit.)
storage temp.   2-8°C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN3261 - class 8 - PG 3 - EHS - Corrosive solid, acidic, organic, n.o.s., HI: all
WGK Germany 
2
RTECS 
BO6822450

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

B4136 Benzyldimethylhexadecylammonium chloride, cationic detergent

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?