• USA Home
  • 13430 - N-Benzylglycine ethyl ester

EMAIL THIS PAGE TO A FRIEND
13430 Sigma-Aldrich

N-Benzylglycine ethyl ester

purum, ≥95.0% (GC)

Synonym: Ethyl (benzylamino)acetate

  • CAS Number 6436-90-4

  • Linear Formula C6H5CH2NHCH2CO2C2H5

  • Molecular Weight 193.24

  •  Beilstein Registry Number 2104816

  •  EC Number 229-218-6

  •  MDL number MFCD00009174

  •  PubChem Substance ID 329750169

Purchase

Properties

InChI Key   ULOLIZHBYWAICY-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
refractive index   n20/D 1.506(lit.)
  n20/D 1.506
bp   140-142 °C/10 mmHg(lit.)
density   1.031 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Reagent used to introduce a protected glycine by N-alkylation

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?